The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167691.1 | NP_001161163.1 | 2 - 3 | 739 | 99 |
NM_133760.1 | NP_598521.1 | 2 - 3 | 647 | 99 | |
GenBank mRNA | AK016400.1 | - | 2 - 3 | 739 | 99 |
BC022653.1 | - | 2 - 3 | 515 | 99 |