Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gtpbp6
Entrez Gene ID
107999
Gene Name
GTP binding protein 6 (putative)
Gene Aliases
AV119224, Pgbpll, pgpl
Chromosome Location
Chr.5: 110103976 - 110108197 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145147.5 NP_660129.2 1 - 2 339 108
GenBank mRNA AK144812.1 - 1 - 2 340 108
AK152225.1 - 1 - 2 340 108
AK152972.1 - 1 - 2 340 108
AK159462.1 - 1 - 2 312 108

View Full Product Details