Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sgms1
Entrez Gene ID
208449
Gene Name
sphingomyelin synthase 1
Gene Aliases
9530058O11Rik, AI841905, C80702, Mob, Sms1, Sor1, Tmem23
Chromosome Location
Chr.19: 32122727 - 32389705 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168526.1 NP_001161998.1 1 - 2 186 87
GenBank mRNA AK079254.1 - 1 - 2 131 87
AK133967.1 - 1 - 2 154 87
AK150272.1 - 1 - 2 164 87
AK152312.1 - 1 - 2 164 87
AK160539.1 - 1 - 2 164 87
BC019443.1 - 1 - 2 153 87

View Full Product Details