Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zdhhc13
Entrez Gene ID
243983
Gene Name
zinc finger, DHHC domain containing 13
Gene Aliases
2410004E01Rik, C530010M18, HIP3RP, Hip14l, kojak, skc4
Chromosome Location
Chr.7: 48789003 - 48827440 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028031.3 NP_082307.1 2 - 3 285 75
GenBank mRNA AK010382.1 - 2 - 3 285 75
AK145855.1 - 2 - 3 205 75
AK146196.1 - 2 - 3 257 75
BC019493.1 - 2 - 3 204 75
BC046599.1 - 2 - 3 204 75

View Full Product Details