Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sgms1
Entrez Gene ID
208449
Gene Name
sphingomyelin synthase 1
Gene Aliases
9530058O11Rik, AI841905, C80702, Mob, Sms1, Sor1, Tmem23
Chromosome Location
Chr.19: 32122727 - 32389705 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144792.4 NP_659041.3 2 - 3 315 84
GenBank mRNA AK082974.1 - 2 - 3 244 84
AK146782.1 - 2 - 3 315 84
AK151170.1 - 2 - 3 300 84
AK153406.1 - 2 - 3 300 84
AY509044.1 - 2 - 3 315 84

View Full Product Details