Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hsd17b13
Entrez Gene ID
243168
Gene Name
hydroxysteroid (17-beta) dehydrogenase 13
Gene Aliases
AI047820, PAN1B-like, Pan1b
Chromosome Location
Chr.5: 103955442 - 103977388 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163486.1 NP_001156958.1 1 - 2 287 71
NM_198030.2 NP_932147.2 1 - 2 287 71
GenBank mRNA AI048624.1 - 1 - 2 274 71
AI573700.1 - 1 - 2 287 71
AY101768.1 - 1 - 2 214 71
BC019427.1 - 1 - 2 263 71

View Full Product Details