Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Saal1
Entrez Gene ID
78935
Gene Name
serum amyloid A-like 1
Gene Aliases
5031425D22Rik, SPACIA1
Chromosome Location
Chr.7: 46686108 - 46710691 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030233.1 NP_084509.1 3 - 4 350 84
GenBank mRNA AB489137.1 - 3 - 4 331 84
AK019878.1 - 3 - 4 351 84
AK030833.1 - 3 - 4 356 84
AK031128.1 - 2 - 3 136 84
AK033202.1 - 3 - 4 361 84
AK035874.1 - 3 - 4 362 84
AK132193.1 - 3 - 4 351 84
AK134418.1 - 3 - 4 358 84
BC019985.1 - 3 - 4 344 84
BC068127.1 - 3 - 4 347 84
BC087944.1 - 3 - 4 362 84

View Full Product Details