The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027340.2 | NP_081616.1 | 1 - 2 | 233 | 89 |
GenBank mRNA | AK008974.1 | - | 1 - 2 | 62 | 89 |
AK154333.1 | - | 1 - 2 | 234 | 89 |