The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029508.3 | NP_083784.1 | 2 - 3 | 366 | 73 |
GenBank mRNA | AK002387.1 | - | 2 - 3 | 389 | 73 |
AK017984.1 | - | 2 - 3 | 367 | 73 | |
AK146133.1 | - | 2 - 3 | 436 | 73 | |
BC139258.1 | - | 2 - 3 | 327 | 73 | |
BC139259.1 | - | 2 - 3 | 327 | 73 | |
BC145590.1 | - | 2 - 3 | 327 | 73 | |
BC145591.1 | - | 2 - 3 | 327 | 73 |