Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sart3
Entrez Gene ID
53890
Gene Name
squamous cell carcinoma antigen recognized by T cells 3
Gene Aliases
AU045857, mKIAA0156
Chromosome Location
Chr.5: 113742444 - 113772505 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016926.1 NP_058622.1 16 - 17 2382 69
GenBank mRNA AF172722.1 - 16 - 17 2382 69
AK086398.1 - 16 - 17 2383 69
BC036350.1 - 15 - 16 2263 69
BC057156.1 - 16 - 17 2373 69

View Full Product Details