Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sart3
Entrez Gene ID
53890
Gene Name
squamous cell carcinoma antigen recognized by T cells 3
Gene Aliases
AU045857, mKIAA0156
Chromosome Location
Chr.5: 113742444 - 113772505 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016926.1 NP_058622.1 3 - 4 565 71
GenBank mRNA AF172722.1 - 3 - 4 565 71
AK053828.1 - 3 - 4 569 71
AK086398.1 - 3 - 4 566 71
AK129067.1 - 3 - 4 560 71
BC036350.1 - 3 - 4 566 71
BC057156.1 - 3 - 4 556 71

View Full Product Details