Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Bcl2l12
- Entrez Gene ID
-
75736
- Gene Name
- BCL2-like 12 (proline rich)
- Gene Aliases
-
2810475P17Rik, 5430429M05Rik, Bcl-L12, Bcl2-L12
- Chromosome Location
-
Chr.7: 44991222 - 44997643 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC039807.1 |
- |
- |
91 |
79 |
View Full Product Details