Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nucb1
Entrez Gene ID
18220
Gene Name
nucleobindin 1
Gene Aliases
B230337F23Rik, C77483, Calnuc, MTEST82, Nucb
Chromosome Location
Chr.7: 45492674 - 45510408 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163662.1 NP_001157134.1 12 - 13 1361 87
NM_008749.2 NP_032775.1 12 - 13 1349 87
GenBank mRNA AK002630.1 - 12 - 13 1342 87
AK004886.1 - 12 - 13 1332 87
AK007976.1 - 12 - 13 1335 87
AK034944.1 - - 56 86
AK045087.1 - 12 - 13 1361 87
AK140710.1 - 12 - 13 1339 87
AK160981.1 - 12 - 13 1348 87
AK164088.1 - 12 - 13 1343 87
AK215214.1 - 1 - 2 43 87
AK216538.1 - 1 - 2 43 87
BC072554.1 - 12 - 13 1339 87
M96823.1 - 11 - 12 1271 87

View Full Product Details