The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195537.1 | NP_001182466.1 | 2 - 3 | 932 | 83 |
GenBank mRNA | AK005822.1 | - | 1 - 2 | 214 | 83 |
BY705974.1 | - | 1 - 2 | 211 | 83 | |
GU144514.1 | - | 2 - 3 | 932 | 83 |