Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Auts2
Entrez Gene ID
319974
Gene Name
autism susceptibility candidate 2
Gene Aliases
2700063G02Rik, A730011F23Rik, D830032G16Rik
Chromosome Location
Chr.5: 131437682 - 132542343 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177047.3 NP_796021.2 6 - 7 1265 90
GenBank mRNA AK147478.1 - - 2264 90
BC096021.1 - 6 - 7 1264 90

View Full Product Details