Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc8d
Entrez Gene ID
231549
Gene Name
leucine rich repeat containing 8D
Gene Aliases
2810473G09Rik, 4930525N13Rik, A930019F03, Lrrc5
Chromosome Location
Chr.5: 105699969 - 105815215 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122768.1 NP_001116240.1 2 - 3 429 132
NM_178701.3 NP_848816.3 2 - 3 419 132
GenBank mRNA AK046249.1 - 2 - 3 428 132
AK050400.1 - 2 - 3 389 132
AK080722.1 - 2 - 3 363 132
AK137459.1 - 2 - 3 430 132
AK167985.1 - 1 - 2 137 132
BC037717.1 - 2 - 3 400 132

View Full Product Details