Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Uhrf2
Entrez Gene ID
109113
Gene Name
ubiquitin-like, containing PHD and RING finger domains 2
Gene Aliases
2310065A22Rik, AI426270, AW214556, D130071B19Rik, Nirf
Chromosome Location
Chr.19: 30030433 - 30093729 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144873.2 NP_659122.2 5 - 6 1243 79
GenBank mRNA AB116653.1 - 5 - 6 978 79
AF274047.1 - 5 - 6 1153 79
AK080925.1 - 5 - 6 1284 79
BC060241.1 - 5 - 6 1243 79

View Full Product Details