Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trpd52l3
Entrez Gene ID
66745
Gene Name
tumor protein D52-like 3
Gene Aliases
4931412G03Rik
Chromosome Location
Chr.19: 30003790 - 30006020 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025741.2 NP_080017.1 1 - 1 2181 72
GenBank mRNA AV265273.1 - 1 - 1 152 72

View Full Product Details