The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163035.1 | NP_001156507.1 | 1 - 2 | 305 | 108 |
NM_026421.3 | NP_080697.2 | 1 - 2 | 305 | 108 | |
GenBank mRNA | AK009954.1 | - | 1 - 2 | 288 | 108 |
AK079062.1 | - | 1 - 2 | 299 | 108 | |
AK080714.1 | - | 1 - 2 | 256 | 108 | |
AK082747.1 | - | 1 - 2 | 280 | 108 | |
AK152021.1 | - | 1 - 2 | 280 | 108 | |
AK159356.1 | - | 1 - 2 | 278 | 108 | |
AK167575.1 | - | 1 - 2 | 316 | 108 | |
AK168696.1 | - | 1 - 2 | 267 | 108 | |
AK177338.1 | - | 1 - 2 | 60 | 108 | |
BC021429.1 | - | 1 - 2 | 232 | 108 |