Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdyhv1
Entrez Gene ID
76773
Gene Name
WDYHV motif containing 1
Gene Aliases
2410187C16Rik, AU014961, AW550036, Ntaq1
Chromosome Location
Chr.15: 58141334 - 58158654 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029734.1 NP_084010.1 5 - 6 580 67
GenBank mRNA AK150578.1 - 5 - 6 632 67
BC051524.1 - 5 - 6 580 67

View Full Product Details