Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snx25
Entrez Gene ID
102141
Gene Name
sorting nexin 25
Gene Aliases
AI661919, Gm1699, Gm173, Sbbi31
Chromosome Location
Chr.8: 46033261 - 46151858 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207213.2 NP_997096.2 8 - 9 1320 130
GenBank mRNA AY601646.1 - 7 - 8 1539 130
BC054725.1 - 3 - 4 576 130
BC057063.1 - 7 - 8 1068 130

View Full Product Details