Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wapal
Entrez Gene ID
218914
Gene Name
wings apart-like homolog (Drosophila)
Gene Aliases
A530089A20Rik, BC037674, DIF-2, FOE, WAPL
Chromosome Location
Chr.14: 34671487 - 34747983 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004436.4 NP_001004436.2 7 - 8 2507 139
NM_001301330.1 NP_001288259.1 7 - 8 2489 139
GenBank mRNA AB167349.1 - 7 - 8 2313 139
AK028167.1 - 5 - 6 611 139
AK129105.1 - 7 - 8 2507 139
BC139434.1 - 7 - 8 2262 139
BC145625.1 - 7 - 8 2244 139

View Full Product Details