Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
N4bp2
Entrez Gene ID
333789
Gene Name
NEDD4 binding protein 2
Gene Aliases
B3bp, E430014I16, Gm1791, Gm868
Chromosome Location
Chr.5: 65761726 - 65830107 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024917.1 NP_001020088.1 15 - 16 4929 140
GenBank mRNA AK136182.1 - 5 - 6 2076 140
AK173167.1 - 13 - 14 4640 140
BC046458.1 - 6 - 7 2909 140
BC054549.1 - 15 - 16 4929 140

View Full Product Details