Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adam18
Entrez Gene ID
13524
Gene Name
a disintegrin and metallopeptidase domain 18
Gene Aliases
Adam27, Dtgn3, Tmdc3, tMDCIII
Chromosome Location
Chr.8: 24602246 - 24674770 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010084.2 NP_034214.2 2 - 3 188 57
GenBank mRNA AF167405.1 - 2 - 3 186 57
AK076633.1 - 2 - 3 189 57
BC051136.1 - 2 - 3 165 57
BC144841.1 - 2 - 3 161 57
BC145751.1 - 2 - 3 161 57
BY099176.1 - 2 - 3 144 57

View Full Product Details