Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam35a
Entrez Gene ID
75698
Gene Name
family with sequence similarity 35, member A
Gene Aliases
3110001K24Rik
Chromosome Location
Chr.14: 34236948 - 34310503 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029389.2 NP_083665.2 6 - 7 2211 112
GenBank mRNA AK081061.1 - 3 - 4 468 112
AK149442.1 - 6 - 7 2075 112
BC144822.1 - 3 - 4 1719 112

View Full Product Details