The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114879.1 | NP_001108351.1 | 21 - 22 | 4316 | 91 |
NM_028945.1 | NP_083221.1 | 21 - 22 | 4316 | 91 | |
GenBank mRNA | AK028620.1 | - | 8 - 9 | 2066 | 91 |
BC037390.1 | - | 7 - 8 | 1807 | 91 | |
BC064458.1 | - | 3 - 4 | 382 | 91 |