The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028746.3 | NP_083022.1 | 3 - 4 | 1255 | 69 |
GenBank mRNA | AB072352.1 | - | 3 - 4 | 1238 | 69 |
AJ417662.1 | - | 3 - 4 | 1204 | 69 | |
AK128901.1 | - | 3 - 4 | 1255 | 69 | |
AK143768.1 | - | 3 - 4 | 1257 | 69 | |
BC014684.1 | - | 3 - 4 | 1228 | 69 |