The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012008.2 | NP_036138.1 | 6 - 7 | 631 | 142 |
GenBank mRNA | AJ007376.1 | - | 6 - 7 | 598 | 142 |
AK154382.1 | - | 6 - 7 | 591 | 142 | |
BC021453.1 | - | 6 - 7 | 596 | 142 | |
BC051046.1 | - | 6 - 7 | 620 | 142 |