Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wwp1
Entrez Gene ID
107568
Gene Name
WW domain containing E3 ubiquitin protein ligase 1
Gene Aliases
8030445B08Rik, AIP5, SDRP1, Tiul1
Chromosome Location
Chr.4: 19608296 - 19709004 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276292.1 NP_001263221.1 16 - 17 1947 112
NM_177327.5 NP_796301.2 17 - 18 2109 112
NR_074089.1 16 - 17 1784 112
NR_074090.1 17 - 18 2006 112
GenBank mRNA AK032232.1 - 16 - 17 1775 112
AK033138.1 - 9 - 10 923 112
AK137588.1 - 17 - 18 2034 112
AK168567.1 - 17 - 18 1973 112
BC026829.1 - 16 - 17 1885 112
BC150812.1 - 16 - 17 1944 112

View Full Product Details