The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001032413.1 | NP_001027585.1 | 10 - 11 | 1328 | 74 |
NM_001032414.1 | NP_001027586.1 | 9 - 10 | 1258 | 74 | |
NM_001289600.1 | NP_001276529.1 | 9 - 10 | 1230 | 74 | |
NM_001289601.1 | NP_001276530.1 | 11 - 12 | 1471 | 74 | |
NM_028460.2 | NP_082736.1 | 11 - 12 | 1462 | 74 | |
NM_152799.2 | NP_690012.2 | 11 - 12 | 1427 | 74 | |
GenBank mRNA | AF440279.1 | - | 11 - 12 | 1452 | 74 |
AF444274.1 | - | 9 - 10 | 1252 | 74 | |
AF461685.1 | - | 8 - 9 | 1185 | 74 | |
AK053551.1 | - | 10 - 11 | 1319 | 74 | |
AK141082.1 | - | - | 1225 | 74 |