Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Casp8ap2
Entrez Gene ID
26885
Gene Name
caspase 8 associated protein 2
Gene Aliases
AA387232, D4Ertd659e, FLASH, mKIAA1315
Chromosome Location
Chr.4: 32615473 - 32653271 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011997.2 NP_036127.2 1 - 2 267 66
GenBank mRNA AF132726.1 - 1 - 2 227 66
BY331503.1 - 1 - 2 263 66

View Full Product Details