The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027491.2 | NP_081767.2 | 1 - 2 | 609 | 112 |
GenBank mRNA | CK387674.1 | - | 1 - 2 | 188 | 112 |