Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clec4g
Entrez Gene ID
75863
Gene Name
C-type lectin domain family 4, member g
Gene Aliases
4930572L20Rik
Chromosome Location
Chr.8: 3707069 - 3721183 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029465.3 NP_083741.1 8 - 9 753 72
GenBank mRNA AK079992.1 - 8 - 9 754 72
BC025069.1 - 8 - 9 745 72
BC087963.1 - 8 - 9 753 72

View Full Product Details