Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pbsn
Entrez Gene ID
54192
Gene Name
probasin
Gene Aliases
PB, Prbs
Chromosome Location
Chr.X: 77837898 - 77853500 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017471.2 NP_059499.1 3 - 4 305 151
GenBank mRNA AF005204.1 - 3 - 4 309 151
AK137501.1 - 3 - 4 332 151

View Full Product Details