The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039466.1 | NP_001034555.1 | 7 - 8 | 1307 | 61 |
NM_183037.2 | NP_898858.1 | 7 - 8 | 1410 | 61 | |
GenBank mRNA | AK132368.1 | - | 7 - 8 | 1277 | 61 |
AK170904.1 | - | 7 - 8 | 1308 | 61 | |
AY251388.1 | - | 7 - 8 | 1290 | 61 |