The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145950.4 | NP_666062.3 | 1 - 2 | 188 | 84 |
GenBank mRNA | AK048731.1 | - | 1 - 2 | 189 | 84 |
AK053737.1 | - | 1 - 2 | 132 | 84 | |
AK148243.1 | - | 1 - 2 | 163 | 84 | |
AK169678.1 | - | 1 - 2 | 133 | 84 | |
CB574822.1 | - | 1 - 2 | 144 | 84 |