Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem33
Entrez Gene ID
67878
Gene Name
transmembrane protein 33
Gene Aliases
1110006G02Rik, 1600019D15Rik, 2410089A21Rik, 2700052H22Rik, 5430406L04Rik, AA388285, AI314185, AI426615, db83
Chromosome Location
Chr.5: 67260565 - 67291461 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285452.1 NP_001272381.1 5 - 6 957 147
NM_028975.4 NP_083251.2 6 - 7 800 147
NM_030108.2 NP_084384.1 6 - 7 800 147
GenBank mRNA AK005493.1 - 6 - 7 692 147
AK007607.1 - 6 - 7 608 147
AK007750.1 - 6 - 7 689 147
AK017281.1 - 6 - 7 669 147
AK032555.1 - 6 - 7 707 147
AK153441.1 - 6 - 7 625 147
AK165369.1 - 6 - 7 602 147
BC005562.1 - 6 - 7 601 147
BC016570.1 - 5 - 6 673 147
BC023966.1 - 5 - 6 666 147

View Full Product Details