Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ido2
Entrez Gene ID
209176
Gene Name
indoleamine 2,3-dioxygenase 2
Gene Aliases
AI265623, C230043N17Rik, Ido-2, Indol1
Chromosome Location
Chr.8: 24531892 - 24576404 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145949.2 NP_666061.3 8 - 9 808 83
GenBank mRNA AK082385.1 - 7 - 8 999 83
EF137182.1 - 8 - 9 808 83

View Full Product Details