Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Kcnj6
- Entrez Gene ID
-
16522
- Gene Name
- potassium inwardly-rectifying channel, subfamily J, member 6
- Gene Aliases
-
BIR1, GIRK2, KATP2, KCNJ7, Kir3.2, weaver, wv
- Chromosome Location
-
Chr.16: 94748683 - 94997696 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB029502.1 |
- |
2 - 3 |
317 |
69 |
View Full Product Details