Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Gnrhr
- Entrez Gene ID
-
14715
- Gene Name
- gonadotropin releasing hormone receptor
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 86181994 - 86197896 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
L28756.1 |
- |
- |
604 |
80 |
View Full Product Details