The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201413.1 | NP_001188342.1 | 8 - 9 | 1757 | 89 |
NM_001201414.1 | NP_001188343.1 | 7 - 8 | 1760 | 89 | |
NM_001201415.1 | NP_001188344.1 | 7 - 8 | 1694 | 89 | |
NM_009686.2 | NP_033816.1 | 8 - 9 | 1757 | 89 | |
GenBank mRNA | AK004792.1 | - | 8 - 9 | 1611 | 89 |
AK167649.1 | - | 8 - 9 | 1622 | 89 | |
AK169959.1 | - | 7 - 8 | 1635 | 89 | |
BC076587.1 | - | 7 - 8 | 1540 | 89 |