Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rbm33
Entrez Gene ID
381626
Gene Name
RNA binding motif protein 33
Gene Aliases
3200001K10Rik, 6430512A10Rik, AA386890, Prr8
Chromosome Location
Chr.5: 28316891 - 28419244 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028234.1 NP_082510.1 5 - 6 738 78
GenBank mRNA AK014289.1 - 5 - 6 727 78

View Full Product Details