Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdcl2
Entrez Gene ID
79455
Gene Name
phosducin-like 2
Gene Aliases
1700010B22Rik, 1700016K07Rik, Mgcphlp, Tphlp
Chromosome Location
Chr.5: 76312127 - 76331172 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK006040.1 - 1 - 2 122 75
AK133140.1 - 1 - 2 83 75

View Full Product Details