Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Usp45
Entrez Gene ID
77593
Gene Name
ubiquitin specific petidase 45
Gene Aliases
3110003C05Rik, 4930550B20Rik, AI843191, Gcap7
Chromosome Location
Chr.4: 21767161 - 21837872 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152825.2 NP_690038.1 12 - 13 1228 141
GenBank mRNA AK019756.1 - 13 - 14 1343 141
BC027768.1 - 12 - 13 1211 141

View Full Product Details