The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026558.4 | NP_080834.3 | 8 - 9 | 1064 | 90 |
GenBank mRNA | AK013129.1 | - | 7 - 8 | 893 | 90 |
AK019574.1 | - | 8 - 9 | 892 | 90 | |
AK078438.1 | - | 8 - 9 | 1045 | 90 | |
AK088705.1 | - | 8 - 9 | 1045 | 90 | |
BC020162.1 | - | - | 858 | 90 | |
BC055848.1 | - | 8 - 9 | 1982 | 90 |