Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myo16
Entrez Gene ID
244281
Gene Name
myosin XVI
Gene Aliases
BM140241, C230040D10Rik, Gm1105, NYAP3, mKIAA0865
Chromosome Location
Chr.8: 10153916 - 10634742 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081397.1 NP_001074866.1 8 - 9 1241 78
GenBank mRNA AB429291.1 - 7 - 8 915 78
AK082350.1 - 8 - 9 977 78
BC072580.1 - 8 - 9 1253 78
BC082311.1 - 8 - 9 1109 78
BC151049.1 - 8 - 9 1195 78
BC151051.1 - 8 - 9 1196 78
BC157966.1 - 8 - 9 1196 78
BC172122.1 - 8 - 9 1196 78

View Full Product Details