Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Thsd1
Entrez Gene ID
56229
Gene Name
thrombospondin, type I, domain 1
Gene Aliases
4833423O18Rik, Tmtsp
Chromosome Location
Chr.8: 22225858 - 22261334 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293688.1 NP_001280617.1 2 - 3 268 103
NM_019576.2 NP_062522.1 2 - 3 268 103
GenBank mRNA AB039946.1 - 1 - 2 96 103
AK049636.1 - 2 - 3 231 103
BC029898.1 - 2 - 3 258 103
BC051957.1 - 2 - 3 226 103
BC119603.2 - 1 - 2 96 103
BY075510.1 - 2 - 3 267 103

View Full Product Details