Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Thsd1
Entrez Gene ID
56229
Gene Name
thrombospondin, type I, domain 1
Gene Aliases
4833423O18Rik, Tmtsp
Chromosome Location
Chr.8: 22225858 - 22261334 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205253.1 NP_001192182.1 1 - 2 124 54
GenBank mRNA AK052555.1 - 1 - 2 65 54

View Full Product Details