The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175013.2 | NP_778178.3 | 3 - 4 | 626 | 73 |
GenBank mRNA | AK035507.1 | - | 3 - 4 | 626 | 73 |
BC115937.1 | - | 3 - 4 | 436 | 73 |