Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zbtb46
Entrez Gene ID
72147
Gene Name
zinc finger and BTB domain containing 46
Gene Aliases
2610019F01Rik, 4933406L05Rik, BZEL, Btbd4
Chromosome Location
Chr.2: 181371261 - 181492982 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028125.3 NP_082401.1 1 - 2 170 95
GenBank mRNA AK011461.1 - 1 - 2 171 95
AK085310.1 - 1 - 2 171 95
AK155148.1 - 1 - 2 148 95
BC058179.1 - 1 - 2 146 95

View Full Product Details